missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphenyl beta-D-thiogalactopyranoside, 98%, Thermo Scientific Chemicals

Catalog No. AAJ6668603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6668603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6668603 Supplier Thermo Scientific Chemicals Supplier No. J6668603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28244-98-6 | Molecular Formula: C13H18O5S | Molecular Weight: 286.342 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, warm methanol, DMSO, or DMF.

Notes
Store at 4°C. Protect from heat. Incompatible with heat and oxidizing agents.

Chemical Identifiers

CAS 28244-98-6
Molecular Formula C13H18O5S
Molecular Weight (g/mol) 286.342
MDL Number MFCD04973631
InChI Key IQCLIQLFPVKINX-SJHCENCUSA-N
Synonym 4-methylphenylthio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-4-methylphenyl sulfanyl oxane-3,4,5-triol, 4-methylphenyl b-d-thiogalactopyranoside, p-tolyl 1-thio-beta-d-galactopyranoside, 4-methylphenyl 1-thio-b-d-galactopyranoside, p-methylphenyl 1-thio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-4-methylphenyl sulfanyloxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-p-tolylthio tetrahydro-2h-pyran-3,4,5-triol
PubChem CID 7315241
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methylphenyl)sulfanyloxane-3,4,5-triol
SMILES CC1=CC=C(C=C1)SC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 137°C to 139°C
Color White
Quantity 1 g
Solubility Information Soluble in water,warm methanol,DMSO,or DMF.
Formula Weight 286.35
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Methylphenyl beta-D-thiogalactopyranoside
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only