missing translation for 'onlineSavingsMsg'
Learn More

(4-Methylphenoxy)acetic acid, 99%, Thermo Scientific™

Catalog No. p-3211022
Change view
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC378270050 5g Glass bottle
AC378270250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC378270050 Supplier Thermo Scientific Chemicals Supplier No. 378270050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 940-64-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00014365
InChI Key SFTDDFBJWUWKMN-UHFFFAOYSA-N
Synonym 4-methylphenoxy acetic acid, 4-methylphenoxyacetic acid, 2-4-methylphenoxy acetic acid, p-methylphenoxyacetic acid, unii-wwy23322ik, acetic acid, 4-methylphenoxy, 2-p-tolyloxy acetic acid, p-toloxyacetic acid, acmc-20apmz, p-tolyloxy-acetic acid
PubChem CID 70329
IUPAC Name 2-(4-methylphenoxy)acetic acid
SMILES CC1=CC=C(C=C1)OCC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3(C6H4)OCH2CO2H
Quantity 5g
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material (4-Methylphenoxy)acetic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-374-7

RUO – Research Use Only