missing translation for 'onlineSavingsMsg'
Learn More

4-Methyloxindole, 95%, Thermo Scientific Chemicals

$190.66 - $591.38

Chemical Identifiers

CAS 13220-46-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD09835631
InChI Key USRZZPHRQZGXFH-UHFFFAOYSA-N
Synonym 4-methylindolin-2-one, 4-methyloxindole, 4-methyl-2-oxo-1,2-dihydro-indole, 2h-indol-2-one, 1,3-dihydro-4-methyl, 4-methylindolinon, 4-methyl-oxindole, 4-methyl-2-oxindole, 4-methyl-1,3-dihydro-indol-2-one, 4-methyl-1,3-dihydro-indole-2-one, 4-methyl-2,3-dihydro-1h-indol-2-one
PubChem CID 12579382
IUPAC Name 4-methyl-2,3-dihydro-1H-indol-2-one
SMILES CC1=C2CC(=O)NC2=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66430MD
View Documents
Thermo Scientific Chemicals
H66430MD
250 mg
Each for $190.66
Only null left
 
AAH6643003
View Documents
Thermo Scientific Chemicals
H6643003
1 g
Each for $591.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13220-46-7
147.18
USRZZPHRQZGXFH-UHFFFAOYSA-N
12579382
CC1=C2CC(=O)NC2=CC=C1
C9H9NO
MFCD09835631
4-methylindolin-2-one, 4-methyloxindole, 4-methyl-2-oxo-1,2-dihydro-indole, 2h-indol-2-one, 1,3-dihydro-4-methyl, 4-methylindolinon, 4-methyl-oxindole, 4-methyl-2-oxindole, 4-methyl-1,3-dihydro-indol-2-one, 4-methyl-1,3-dihydro-indole-2-one, 4-methyl-2,3-dihydro-1h-indol-2-one
4-methyl-2,3-dihydro-1H-indol-2-one

Specifications

13220-46-7
MFCD09835631
119249
USRZZPHRQZGXFH-UHFFFAOYSA-N
4-methyl-2,3-dihydro-1H-indol-2-one
12579382
95%
4-Methyloxindole
C9H9NO
250 mg
4-methylindolin-2-one, 4-methyloxindole, 4-methyl-2-oxo-1,2-dihydro-indole, 2h-indol-2-one, 1,3-dihydro-4-methyl, 4-methylindolinon, 4-methyl-oxindole, 4-methyl-2-oxindole, 4-methyl-1,3-dihydro-indol-2-one, 4-methyl-1,3-dihydro-indole-2-one, 4-methyl-2,3-dihydro-1h-indol-2-one
CC1=C2CC(=O)NC2=CC=C1
147.18
147.18
Solid
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only