missing translation for 'onlineSavingsMsg'
Learn More

4-Methylnaphthalene-1-boronic acid, 96%

Catalog No. AAH5299803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5299803 1 g
AAH5299806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5299803 Supplier Thermo Scientific Chemicals Supplier No. H5299803

CAS: 103986-53-4 | C11H11BO2 | 186.02 g/mol

Reactant for Suzuki-Miyaura coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for Suzuki-Miyaura coupling.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 103986-53-4
Molecular Formula C11H11BO2
Molecular Weight (g/mol) 186.02
MDL Number MFCD01632204
InChI Key JHVQEUGNYSVSDH-UHFFFAOYSA-N
Synonym 4-methyl-1-naphthaleneboronic acid, 4-methyl-1-naphthalene boronic acid, 4-methylnaphthalene-1-boronic acid, 1-methylnaphthalene-4-boronic acid, 4-methylnaphthalen-1-yl boronic acid, 1-borono-4-methylnaphthalene, 4-methyl-1-naphthyl boronic acid, 4-methyl-1-naphthylboronic acid, 4-methylnaphthalen-1-yl boranediol, boronic acid, 4-methyl-1-naphthalenyl
PubChem CID 2773511
IUPAC Name (4-methylnaphthalen-1-yl)boronic acid
SMILES CC1=CC=C(B(O)O)C2=CC=CC=C12

Specifications

Melting Point 239°C to 240°C
Quantity 1 g
Formula Weight 186.02
Percent Purity 96%
Chemical Name or Material 4-Methylnaphthalene-1-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only