missing translation for 'onlineSavingsMsg'
Learn More

4-Methylmorpholine 99.0+%, TCI America™

Catalog No. M0370500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0370500ML 500 mL
M037025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0370500ML Supplier TCI America Supplier No. M0370500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-02-4
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00006175
InChI Key SJRJJKPEHAURKC-UHFFFAOYSA-N
Synonym n-methylmorpholine, morpholine, 4-methyl, methylmorpholine, 1-methylmorpholine, morpholine, n-methyl, 4-methylmorfolin, 4-methylmorpholin, n-methyl morpholine, 4-methylmorfolin czech, n-methylmorpholin
PubChem CID 7972
IUPAC Name 4-methylmorpholine
SMILES CN1CCOCC1

Specifications

Melting Point -66°C
Color Yellow
Boiling Point 116°C
Quantity 500 mL
UN Number 2535
Formula Weight 101.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylmorpholine
EINECSNumber (5)-0860
RTECSNumber QE5775000
TSCA Yes