missing translation for 'onlineSavingsMsg'
Learn More

4-Methylmorpholine N-Oxide 98.0+%, TCI America™

Catalog No. p-7133082
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
M21925G 5 g
M219225G 25 g
2 options

Catalog No. M21925G

Supplier: TCI America M21925G

Only null left

Chemical Identifiers

CAS 7529-22-8
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00005947
InChI Key LFTLOKWAGJYHHR-UHFFFAOYSA-N
Synonym 4-methylmorpholine n-oxide, 4-methylmorpholine 4-oxide, n-methylmorpholine oxide, n-methylmorpholine n-oxide, morpholine, 4-methyl-, 4-oxide, 4-methylmorpholine-4-oxide, methyl morpholine oxide, n-methylmorpholine-n-oxide, nmo, unii-arc64pkj0f
PubChem CID 82029
ChEBI CHEBI:52093
IUPAC Name 4-methylmorpholin-4-ium-4-olate
SMILES C[N+]1([O-])CCOCC1

Specifications

Melting Point 177°C
Quantity 5 g
Formula Weight 117.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylmorpholine N-Oxide
TSCA Yes
Recommended Storage Refrigerator