missing translation for 'onlineSavingsMsg'
Learn More

4-Methylmorpholine N-oxide monohydrate, 98+%

Catalog No. AAA1599622
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1599622 100 g
AAA1599609 10 g
AAA1599614 25 g
AAA1599618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1599622 Supplier Thermo Scientific Chemicals Supplier No. A1599622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 70187-32-5 | C5H13NO3 | 135.163 g/mol

4-Methylmorpholine N-oxide monohydrate is used as a solvent to prepare cellulose fibers. It is an oxidant and involved in the catalytic OsO4 oxidation of olefins to cis-1,2-diols. It is also involved in ruthenium catalyzed oxidation of alcohols to aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylmorpholine N-oxide monohydrate is used as a solvent to prepare cellulose fibers. It is an oxidant and involved in the catalytic OsO4 oxidation of olefins to cis-1,2-diols. It is also involved in ruthenium catalyzed oxidation of alcohols to aldehydes and ketones.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool place. Keep the container tightly closed. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 70187-32-5
Molecular Formula C5H13NO3
Molecular Weight (g/mol) 135.163
MDL Number MFCD00149388
InChI Key WAZPLXZGZWWXDQ-UHFFFAOYSA-N
Synonym 4-methylmorpholine n-oxide monohydrate, 4-methylmorpholine 4-oxide hydrate, 4-methylmorpholine n-oxide hydrate, 4-methylmorpholine 4-oxide xhydrate, unii-087wu2ftq5, n-methylmorpholine n-oxide monohydrate, n-methylmorpholine n-oxide hydrate, n-methylmorpholine-n-oxide monohydrate, 4-methyl-4-oxidomorpholin-4-ium hydrate, morpholine, 4-methyl-, 4-oxide, hydrate
PubChem CID 2724197
IUPAC Name 4-methyl-4-oxidomorpholin-4-ium;hydrate
SMILES C[N+]1(CCOCC1)[O-].O

Specifications

Melting Point 73°C to 76°C
Quantity 100 g
Beilstein 5449441
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 135.17 (117.15 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 4-Methylmorpholine N-oxide monohydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-391-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only