missing translation for 'onlineSavingsMsg'
Learn More

4-Methylmorpholine N-Oxide (50% in Water, ca. 4.8mol/L), TCI America™

Catalog No. M098125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M098125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M098125ML Supplier TCI America Supplier No. M098125ML

Chemical Identifiers

CAS 7529-22-8
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00005947
InChI Key LFTLOKWAGJYHHR-UHFFFAOYSA-N
Synonym 4-methylmorpholine n-oxide, 4-methylmorpholine 4-oxide, n-methylmorpholine oxide, n-methylmorpholine n-oxide, morpholine, 4-methyl-, 4-oxide, 4-methylmorpholine-4-oxide, methyl morpholine oxide, n-methylmorpholine-n-oxide, nmo, unii-arc64pkj0f
PubChem CID 82029
ChEBI CHEBI:52093
IUPAC Name 4-methylmorpholin-4-ium-4-olate
SMILES C[N+]1([O-])CCOCC1

Specifications

Color Yellow
Boiling Point 119°C
Quantity 25 mL
Formula Weight 117.15
Physical Form Liquid
Chemical Name or Material 4-Methylmorpholine N-Oxide (50% in Water, ca. 4.8mol/L)