missing translation for 'onlineSavingsMsg'
Learn More

4-Methylindole 98.0+%, TCI America™

Catalog No. M17451G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M17451G Supplier TCI America Supplier No. M17451G

Chemical Identifiers

CAS 16096-32-5
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005668
InChI Key PZOUSPYUWWUPPK-UHFFFAOYSA-N
Synonym 4-methylindole, 4-methyindole, 1h-indole, 4-methyl, indole, 4-methyl, 4-methyl indole, 4-methyl-indole, pubchem7233, acmc-1bpu3, ksc179o0l
PubChem CID 85282
IUPAC Name 4-methyl-1H-indole
SMILES CC1=C2C=CNC2=CC=C1

Specifications

Melting Point 5°C
Color Yellow
Boiling Point 267°C
Quantity 1 g
Formula Weight 131.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylindole
TSCA No
Recommended Storage Refrigerator