missing translation for 'onlineSavingsMsg'
Learn More

4-Methylindole, 99%

Catalog No. p-164125
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC133890010 1 g Glass bottle
AC133890050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC133890010 Supplier Thermo Scientific Chemicals Supplier No. 133890010

CAS: 16096-32-5 | C9H9N | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16096-32-5
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00005668
InChI Key PZOUSPYUWWUPPK-UHFFFAOYSA-N
Synonym 4-methylindole, 4-methyindole, 1h-indole, 4-methyl, indole, 4-methyl, 4-methyl indole, 4-methyl-indole, pubchem7233, acmc-1bpu3, ksc179o0l
PubChem CID 85282
IUPAC Name 4-methyl-1H-indole
SMILES CC1=C2C=CNC2=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 5°C
Color Yellow to Brown
Density 1.0600g/mL
Boiling Point 267°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6060 to 1.608
Quantity 1 g
Beilstein 20,II,204
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether and acetone
Formula Weight 131.18
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material 4-Methylindole, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 240-262-5
Recommended Storage This product darkens on exposure to light

RUO – Research Use Only