missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylimidazole 98.0+%, TCI America™
$70.84 - $425.00
Chemical Identifiers
| CAS | 822-36-6 |
|---|---|
| Molecular Formula | C4H6N2 |
| Molecular Weight (g/mol) | 82.11 |
| MDL Number | MFCD00005201 |
| InChI Key | XLSZMDLNRCVEIJ-UHFFFAOYSA-N |
| Synonym | 4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol |
| PubChem CID | 13195 |
| ChEBI | CHEBI:40035 |
| IUPAC Name | 5-methyl-1H-imidazole |
| SMILES | CC1=CN=CN1 |
Chemical Identifiers
| 822-36-6 | |
| 82.11 | |
| XLSZMDLNRCVEIJ-UHFFFAOYSA-N | |
| 13195 | |
| 5-methyl-1H-imidazole |
| C4H6N2 | |
| MFCD00005201 | |
| 4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol | |
| CHEBI:40035 | |
| CC1=CN=CN1 |
Specifications
| 822-36-6 | |
| Yellow | |
| C4H6N2 | |
| 25 g | |
| 4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol | |
| CC1=CN=CN1 | |
| 82.11 | |
| CHEBI:40035 | |
| ≥98.0% (GC) | |
| 4-Methylimidazole |
| 52°C | |
| 263°C | |
| MFCD00005201 | |
| 3263 | |
| XLSZMDLNRCVEIJ-UHFFFAOYSA-N | |
| 5-methyl-1H-imidazole | |
| 13195 | |
| 82.11 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-5709
RTECSNumber : NI7350000
TSCA : Yes