missing translation for 'onlineSavingsMsg'
Learn More

4-Methylimidazole 98.0+%, TCI America™

Catalog No. M0636500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0636500G 500 g
M063625G 25 g
M0636100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0636500G Supplier TCI America Supplier No. M0636500G

Chemical Identifiers

CAS 822-36-6
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005201
InChI Key XLSZMDLNRCVEIJ-UHFFFAOYSA-N
Synonym 4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
PubChem CID 13195
ChEBI CHEBI:40035
IUPAC Name 5-methyl-1H-imidazole
SMILES CC1=CN=CN1

Specifications

Melting Point 52°C
Color Yellow
Boiling Point 263°C
Quantity 500 g
UN Number 3263
Formula Weight 82.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylimidazole
EINECSNumber (5)-5709
RTECSNumber NI7350000
TSCA Yes