missing translation for 'onlineSavingsMsg'
Learn More

4-Methylimidazole, 98%

CAS: 822-36-6 | C4H6N2 | 82.11 g/mol

$53.48 - $344.23

Chemical Identifiers

CAS 822-36-6
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005201
InChI Key XLSZMDLNRCVEIJ-UHFFFAOYSA-N
Synonym 4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
PubChem CID 13195
ChEBI CHEBI:40035
IUPAC Name 5-methyl-1H-imidazole
SMILES CC1=CN=CN1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2413909
View Documents
Thermo Scientific Chemicals
B2413909
10 g
Each for $53.48
Only null left
 
AAB2413918
View Documents
Thermo Scientific Chemicals
B2413918
50 g
Each for $96.93
Only null left
 
AAB2413930
View Documents
Thermo Scientific Chemicals
B2413930
250 g
Each for $344.23
Only null left
 
Description

Description

4-Methylimidazole is used for the curing agent of epoxy resin, the main raw material of medicine cimetidine. It can also be used for synthesis of antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylimidazole is used for the curing agent of epoxy resin, the main raw material of medicine cimetidine. It can also be used for synthesis of antibacterial agent.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Specifications

Chemical Identifiers

822-36-6
82.11
XLSZMDLNRCVEIJ-UHFFFAOYSA-N
13195
5-methyl-1H-imidazole
C4H6N2
MFCD00005201
4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
CHEBI:40035
CC1=CN=CN1

Specifications

822-36-6
1.02
157°C (314°F)
MFCD00005201
UN3263
4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
XLSZMDLNRCVEIJ-UHFFFAOYSA-N
5-methyl-1H-imidazole
13195
82.11
4-Methylimidazole
49°C to 57°C
263°C
C4H6N2
10 g
1453
Soluble in water.
CC1=CN=CN1
82.11
CHEBI:40035
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H311-H314-H335-H350

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 212-497-3

RTECSNumber : NI7350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only