missing translation for 'onlineSavingsMsg'
Learn More

4-Methylimidazole, 98%

CAS: 822-36-6 | C4H6N2 | 82.11 g/mol

$102.90 - $102.90

Chemical Identifiers

CAS 822-36-6
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005201
InChI Key XLSZMDLNRCVEIJ-UHFFFAOYSA-N
Synonym 4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
PubChem CID 13195
ChEBI CHEBI:40035
IUPAC Name 5-methyl-1H-imidazole
SMILES CC1=CN=CN1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC190330500
SDS View Documents
Thermo Scientific Chemicals
190330500
50 g Glass bottle
Each for $102.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

822-36-6
82.11
XLSZMDLNRCVEIJ-UHFFFAOYSA-N
13195
5-methyl-1H-imidazole
C4H6N2
MFCD00005201
4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
CHEBI:40035
CC1=CN=CN1

Specifications

822-36-6
100.0
Brown to Brown-Red
157°C
97.5% min. (GC)
C4H6N2
50 g
4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
XLSZMDLNRCVEIJ-UHFFFAOYSA-N
5-methyl-1H-imidazole
13195
82.11
Powder
97.5
52.0°C to 56.0°C
263.0°C
Authentic
Glass bottle
MFCD00005201
23, 69
Solubility in water: soluble. Other solubilities: soluble in almost all common organic solvents
CC1=CN=CN1
82.11
CHEBI:40035
98%
4-Methylimidazole, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 212-497-3

RTECSNumber : NI7350000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only