missing translation for 'onlineSavingsMsg'
Learn More

4-Methylimidazole, 98%

Catalog No. AAB2413930
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2413930 250 g
AAB2413909 10 g
AAB2413918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2413930 Supplier Thermo Scientific Chemicals Supplier No. B2413930
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 822-36-6 | C4H6N2 | 82.11 g/mol

4-Methylimidazole is used for the curing agent of epoxy resin, the main raw material of medicine cimetidine. It can also be used for synthesis of antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylimidazole is used for the curing agent of epoxy resin, the main raw material of medicine cimetidine. It can also be used for synthesis of antibacterial agent.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.

Chemical Identifiers

CAS 822-36-6
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005201
InChI Key XLSZMDLNRCVEIJ-UHFFFAOYSA-N
Synonym 4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
PubChem CID 13195
ChEBI CHEBI:40035
IUPAC Name 5-methyl-1H-imidazole
SMILES CC1=CN=CN1

Specifications

Melting Point 49°C to 57°C
Density 1.02
Boiling Point 263°C
Flash Point 157°C (314°F)
Quantity 250 g
UN Number UN3263
Beilstein 1453
Solubility Information Soluble in water.
Formula Weight 82.11
Percent Purity 98%
Chemical Name or Material 4-Methylimidazole
Show More Show Less
Hazard Category H302-H311-H314-H335-H350
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 212-497-3
RTECSNumber NI7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only