missing translation for 'onlineSavingsMsg'
Learn More

4-Methylhomopiperazine-4-aminobenzene trihydrochloride monohydrate, 90%, Thermo Scientific™

Chemical Identifiers

CAS 913830-33-8
Molecular Formula C12H24Cl3N3O
Molecular Weight (g/mol) 332.694
MDL Number MFCD09025902
InChI Key RSGOMRQTIZQMIR-UHFFFAOYSA-N
Synonym 4-methylhomopiperazine-4-aminobenzene trihydrochloride monohydrate, 4-4-methyl-1,4-diazepan-1-yl aniline hydrate trihydrochloride, 4-4-methyl-1,4-diazepan-1-yl aniline trihydrochloride hydrate, 1-4-aminophenyl-4-methylhomopiperazine trihydrochloride hydrate, 4-4-methylhomopiperazin-1-yl aniline trihydrochloride monohydrate, 4-4-methyl-1,4-diazepan-1-yl aniline-hydrogen chloride-water 1/3/1
PubChem CID 44119580
IUPAC Name 4-(4-methyl-1,4-diazepan-1-yl)aniline;hydrate;trihydrochloride
SMILES CN1CCCN(CC1)C2=CC=C(C=C2)N.O.Cl.Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC60364CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC60364DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

913830-33-8
332.694
RSGOMRQTIZQMIR-UHFFFAOYSA-N
44119580
CN1CCCN(CC1)C2=CC=C(C=C2)N.O.Cl.Cl.Cl
C12H24Cl3N3O
MFCD09025902
4-methylhomopiperazine-4-aminobenzene trihydrochloride monohydrate, 4-4-methyl-1,4-diazepan-1-yl aniline hydrate trihydrochloride, 4-4-methyl-1,4-diazepan-1-yl aniline trihydrochloride hydrate, 1-4-aminophenyl-4-methylhomopiperazine trihydrochloride hydrate, 4-4-methylhomopiperazin-1-yl aniline trihydrochloride monohydrate, 4-4-methyl-1,4-diazepan-1-yl aniline-hydrogen chloride-water 1/3/1
4-(4-methyl-1,4-diazepan-1-yl)aniline;hydrate;trihydrochloride

Specifications

913830-33-8
100.0
C12H24Cl3N3O
MFCD09025902
RSGOMRQTIZQMIR-UHFFFAOYSA-N
4-(4-methyl-1,4-diazepan-1-yl)aniline;hydrate;trihydrochloride
44119580
90%
89.5
90%
Yes
4-methylhomopiperazine-4-aminobenzene trihydrochloride monohydrate, 4-4-methyl-1,4-diazepan-1-yl aniline hydrate trihydrochloride, 4-4-methyl-1,4-diazepan-1-yl aniline trihydrochloride hydrate, 1-4-aminophenyl-4-methylhomopiperazine trihydrochloride hydrate, 4-4-methylhomopiperazin-1-yl aniline trihydrochloride monohydrate, 4-4-methyl-1,4-diazepan-1-yl aniline-hydrogen chloride-water 1/3/1
CN1CCCN(CC1)C2=CC=C(C=C2)N.O.Cl.Cl.Cl
332.694
332.71
4-Methylhomopiperazine-4-aminobenzene trihydrochloride monohydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents