missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexylamine (cis- and trans- mixture) 99.0+%, TCI America™

Catalog No. M11175ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11175ML 5 mL
M111725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11175ML Supplier TCI America Supplier No. M11175ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6321-23-9
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD06411232,MFCD00001495,MFCD13563117
InChI Key KSMVBYPXNKCPAJ-UHFFFAOYSA-N
Synonym 4-methylcyclohexylamine, trans-4-methylcyclohexylamine, 4-methylcyclohexanamine, trans-4-methylcyclohexanamine, cyclohexanamine, 4-methyl, cis-4-methylcyclohexylamine, p-methylcyclohexylamine, trans-4-methyl-cyclohexylamine, trans-4-methylcyclohexyl amine, 4-methyl-cyclohexylamine
PubChem CID 80604
IUPAC Name 4-methylcyclohexan-1-amine
SMILES CC1CCC(N)CC1

Specifications

Color Yellow
Boiling Point 152°C
Quantity 5 mL
UN Number 2734
Formula Weight 113.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylcyclohexylamine (cis- and trans- mixture)