missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexanone, 98+%

Catalog No. AAA1318622
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1318622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1318622 Supplier Thermo Scientific Chemicals Supplier No. A1318622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 589-92-4 | C7H12O | 112.17 g/mol

4-Methylcyclohexanone was used in the synthesis of the pinguisane-type sesquiterpenoid acutifolone A, in racemic form. It is used as intermediates of CCNU and it can be also be used as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylcyclohexanone was used in the synthesis of the pinguisane-type sesquiterpenoid acutifolone A, in racemic form. It is used as intermediates of CCNU and it can be also be used as a solvent.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 589-92-4
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00001643
InChI Key VGVHNLRUAMRIEW-UHFFFAOYSA-N
Synonym 4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french
PubChem CID 11525
IUPAC Name 4-methylcyclohexan-1-one
SMILES CC1CCC(=O)CC1

Specifications

Melting Point -41°C
Density 0.914
Boiling Point 169°C to 171°C
Flash Point 48°C (118°F)
Refractive Index 1.445
Quantity 100 g
UN Number UN2297
Beilstein 506746
Solubility Information Not miscible in water.
Formula Weight 112.17
Percent Purity ≥98%
Chemical Name or Material 4-Methylcyclohexanone
Show More Show Less
Hazard Category H226-H302+H312+H332
Hazard Statement GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYLCYCLOHEXANONE
EINECSNumber 209-665-3
RTECSNumber GW1750200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only