missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7132777
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16215G 5 g
M162125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16215G Supplier TCI America Supplier No. M16215G

Chemical Identifiers

CAS 4331-54-8
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00074943,MFCD00074909,MFCD20617670
InChI Key QTDXSEZXAPHVBI-UHFFFAOYSA-N
PubChem CID 20330
IUPAC Name 4-methylcyclohexane-1-carboxylic acid
SMILES CC1CCC(CC1)C(O)=O

Specifications

Color Colorless
Boiling Point 140°C
Quantity 5 g
Formula Weight 142.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Methylcyclohexanecarboxylic Acid (cis- and trans- mixture)
EINECSNumber (3)-3311
TSCA No