missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexane-1,2-dicarboxylic Anhydride 98.0+%, TCI America™

Catalog No. p-7132243
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M056725G 25 g
M0567500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M056725G Supplier TCI America Supplier No. M056725G
Only null left

Chemical Identifiers

CAS 19438-60-9
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00005927
InChI Key FKBMTBAXDISZGN-UHFFFAOYSA-N
Synonym hexahydro-4-methylphthalic anhydride, 4-methylhexahydrophthalic anhydride, 5-methylhexahydroisobenzofuran-1,3-dione, 4-methyl-1,2-cyclohexanedicarboxylic anhydride, 1,3-isobenzofurandione, hexahydro-5-methyl, 1,3-isobenzofurandione, hexahydromethyl, 4-methylcyclohexane-1,2-dicarboxylic anhydride, 5-methylhexahydrophthalic anhydride, 5-methyl-3a,4,5,6,7,7a-hexahydroisobenzofuran-1,3-dione, 4-methyl-1,2-cyclohexanedicarboxylic anhydride, mixture of isomers
PubChem CID 86876
IUPAC Name 5-methyl-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES CC1CCC2C(C1)C(=O)OC2=O

Specifications

Color Colorless
Quantity 25 g
Formula Weight 168.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylcyclohexane-1,2-dicarboxylic Anhydride
EINECSNumber (3)-2450
TSCA Yes