missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylbiphenyl-4-carboxaldehyde, 96%

Catalog No. AAH34249MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34249MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH34249MD Supplier Thermo Scientific Chemicals Supplier No. H34249MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 36393-42-7 | C14H12O | 196.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36393-42-7
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD02258938
InChI Key BCINBWXQYBLSKO-UHFFFAOYSA-N
Synonym 4-4-methylphenyl benzaldehyde, 4'-methyl-1,1'-biphenyl-4-carbaldehyde, 4'-methylbiphenyl-4-carbaldehyde, 4'-methyl-4-formylbiphenyl, 1,1'-biphenyl-4-carboxaldehyde, 4'-methyl, 4'-methyl-biphenyl-4-carbaldehyde, 4'-methylbiphenyl-4-carboxaldehyde, 4'-methyl-biphenyl-4-carboxaldehyde, 4'-methyl 1,1'-biphenyl-4-carbaldehyde, pubchem10296
PubChem CID 1392762
IUPAC Name 4-(4-methylphenyl)benzaldehyde
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)C=O

Specifications

Melting Point 108°C to 113°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 196.25
Percent Purity 96%
Chemical Name or Material 4'-Methylbiphenyl-4-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only