Learn More
4-Methylbenzyl cyanide, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 2947-61-7 |
|---|---|
| Molecular Formula | C9H9N |
| Molecular Weight (g/mol) | 131.18 |
| MDL Number | MFCD00001922 |
| InChI Key | RNHKXHKUKJXLAU-UHFFFAOYSA-N |
| Synonym | 4-methylbenzyl cyanide, 4-methylphenylacetonitrile, p-tolylacetonitrile, p-methylbenzyl cyanide, 2-4-methylphenyl acetonitrile, benzeneacetonitrile, 4-methyl, 4-methylphenyl acetonitrile, 4-methylbenzeneacetonitrile, 4-methylbenzylcyanide, 2-p-tolylacetonitrile |
| PubChem CID | 76280 |
| IUPAC Name | 2-(4-methylphenyl)acetonitrile |
| SMILES | CC1=CC=C(CC#N)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC126440050
|
Thermo Scientific Chemicals
126440050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC126440250
|
Thermo Scientific Chemicals
126440250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC126441000
|
Thermo Scientific Chemicals
126441000 |
100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 2947-61-7 | |
| 131.18 | |
| RNHKXHKUKJXLAU-UHFFFAOYSA-N | |
| 76280 | |
| CC1=CC=C(CC#N)C=C1 |
| C9H9N | |
| MFCD00001922 | |
| 4-methylbenzyl cyanide, 4-methylphenylacetonitrile, p-tolylacetonitrile, p-methylbenzyl cyanide, 2-4-methylphenyl acetonitrile, benzeneacetonitrile, 4-methyl, 4-methylphenyl acetonitrile, 4-methylbenzeneacetonitrile, 4-methylbenzylcyanide, 2-p-tolylacetonitrile | |
| 2-(4-methylphenyl)acetonitrile |
Specifications
| 2947-61-7 | |
| 100.0 | |
| 242°C to 243°C | |
| Authentic | |
| Glass bottle | |
| 1.5170 to 1.5195 | |
| MFCD00001922 | |
| 09, 530 | |
| 4-methylbenzyl cyanide, 4-methylphenylacetonitrile, p-tolylacetonitrile, p-methylbenzyl cyanide, 2-4-methylphenyl acetonitrile, benzeneacetonitrile, 4-methyl, 4-methylphenyl acetonitrile, 4-methylbenzeneacetonitrile, 4-methylbenzylcyanide, 2-p-tolylacetonitrile | |
| RNHKXHKUKJXLAU-UHFFFAOYSA-N | |
| 2-(4-methylphenyl)acetonitrile | |
| 76280 | |
| 98% |
| 97.5 | |
| 0.9900g/mL | |
| 106°C | |
| 97% min. (GC) | |
| C9H9N | |
| CH3C6H4CH2CN | |
| 5g | |
| 0.99 | |
| Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether,benzene | |
| CC1=CC=C(CC#N)C=C1 | |
| 131.18 | |
| 131.18 | |
| 4-Methylbenzyl cyanide |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 220-963-2
RUO â Research Use Only