missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzamidine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7133384
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27141G 1 g
M27145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27141G Supplier TCI America Supplier No. M27141G

Chemical Identifiers

CAS 6326-27-8
Molecular Formula C8H11ClN2
Molecular Weight (g/mol) 170.64
MDL Number MFCD00040183
InChI Key MTDUPZAXNYELOU-UHFFFAOYSA-N
PubChem CID 122753
IUPAC Name 4-methylbenzenecarboximidamide;hydrochloride
SMILES CC1=CC=C(C=C1)C(=N)N.Cl

Specifications

Melting Point 215°C
Color White
Quantity 1 g
Formula Weight 170.64
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylbenzamidine Hydrochloride
RTECSNumber XS4865000
TSCA No