missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)pyridine 98.0+%, TCI America™

Catalog No. M21831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21831G 1 g
M21835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21831G Supplier TCI America Supplier No. M21831G

Chemical Identifiers

CAS 1121-58-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00152625
InChI Key LSCYTCMNCWMCQE-UHFFFAOYSA-N
Synonym 4-methylamino pyridine, 4-methylaminopyridine, n-methyl-4-pyridinamine, 4-pyridinamine, n-methyl, pyridin-4-yl methyl amine, 4-mamp, dmap on polystyrene, pubchem12915, pyridin-4yl-methylamine, pyridin-4-yl-methylamine
PubChem CID 123098
IUPAC Name N-methylpyridin-4-amine
SMILES CNC1=CC=NC=C1

Specifications

Melting Point 127°C
Color White-Yellow
Boiling Point 100°C
Quantity 1 g
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylamino)pyridine