missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)benzonitrile, 97%

Catalog No. p-7041709
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6514806 5 g
AAH6514814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6514806 Supplier Thermo Scientific Chemicals Supplier No. H6514806
Only null left

CAS: 4714-62-9 | C8H8N2 | 132.166 g/mol

4-(Methylamino)benzonitrile is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylamino)benzonitrile is used as pharmaceutical intermediates.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 4714-62-9
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.166
MDL Number MFCD00016381
InChI Key JEDXDYXIRZOEKG-UHFFFAOYSA-N
Synonym 4-methylamino benzonitrile, 4-n-methylamino benzonitrile, 4-cyano-n-methylaniline, 4-methylamino-benzonitrile, benzonitrile, 4-methylamino, pubchem4815, 4-cyano-n-methyl-aniline, ksc497o2f, 4-n-methyl aminobenzonitrile, 4-n-methylamino-benzonitrile
PubChem CID 10964551
IUPAC Name 4-(methylamino)benzonitrile
SMILES CNC1=CC=C(C=C1)C#N

Specifications

Melting Point 87°C to 91°C
Quantity 5 g
Beilstein 2802435
Solubility Information Soluble in methanol.
Formula Weight 132.17
Percent Purity 97%
Chemical Name or Material 4-(Methylamino)benzonitrile
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only