missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylacetoacetanilide 98.0+%, TCI America™

Catalog No. A102125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A102125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A102125G Supplier TCI America Supplier No. A102125G

Specifications

CAS 2415-85-2
Melting Point 95°C
Color White-Yellow
Molecular Formula C11H13NO2
Quantity 25 g
MDL Number MFCD00043931
UN Number 2811
Synonym N-Acetoacetyl-4-methylaniline, N-Acetoacetyl-p-toluidine
InChI Key MJGLMEMIYDUEHA-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)NC(=O)CC(=O)C
IUPAC Name N-(4-methylphenyl)-3-oxobutanamide
Molecular Weight (g/mol) 191.23
PubChem CID 17015
Formula Weight 191.23
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Methylacetoacetanilide
Show More Show Less
EINECSNumber (3)-0204
TSCA Yes