missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific™

Catalog No. CC13539CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13539CB 250 mg Amber Glass Bottle
CC13539DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC13539CB Supplier Thermo Scientific Chemicals Supplier No. CC13539CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 519054-54-7
Molecular Formula C15H22BNO3
Molecular Weight (g/mol) 275.16
MDL Number MFCD04115377
InChI Key QRAOZQGIUIDZQZ-UHFFFAOYSA-N
Synonym 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 4-methyl-3,4-dihydro-2h-1,4-benzoxazin-7-ylboronic acid pinacol ester, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1,4-benzoxazine, 4-methyl-7-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo 1,4 oxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2,-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-d ioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine
PubChem CID 2795301
IUPAC Name 4-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1,4-benzoxazine
SMILES CN1CCOC2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 275.15
Chemical Name or Material 4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine