missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-6,7-dihydro-4H-thieno[3,2-c]pyran-4-yl)acetic acid, 97%, Thermo Scientific™

Catalog No. p-3541080
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC61801CB 250mg Amber Glass Bottle
CC61801DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC61801CB Supplier Thermo Scientific Chemicals Supplier No. CC61801CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57153-43-2
Molecular Formula C10H12O3S
Molecular Weight (g/mol) 212.263
MDL Number MFCD01850960
InChI Key MBZWBAMMUGJINB-UHFFFAOYSA-N
Synonym 4-methyl-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl acetic acid, 4-methyl-6h,7h-thieno 3,2-c pyran-4-yl acetic acid, 4h-thieno 3,2-c pyran-4-aceticacid, 6,7-dihydro-4-methyl, 2-4-methyl-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl acetic acid, 2-4-methyl-6,7-dihydro-4h-thiopheno 3,2-c pyran-4-yl acetic acid
PubChem CID 12225898
IUPAC Name 2-(4-methyl-6,7-dihydrothieno[3,2-c]pyran-4-yl)acetic acid
SMILES CC1(C2=C(CCO1)SC=C2)CC(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 212.27
Percent Purity 97%
Chemical Name or Material (4-Methyl-6, 7-dihydro-4H-thieno[3, 2-c]pyran-4-yl)acetic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.