missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-5-vinylthiazole 96.0+%, TCI America™

Catalog No. M10295G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10295G Supplier TCI America Supplier No. M10295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1759-28-0
Molecular Formula C6H7NS
Molecular Weight (g/mol) 125.19
MDL Number MFCD00005337
InChI Key QUAMMXIRDIIGDJ-UHFFFAOYSA-N
Synonym 4-methyl-5-vinylthiazole, thiazole, 5-ethenyl-4-methyl, 4-methyl-5-vinyl thiazole, 5-ethenyl-4-methylthiazole, thiazole, 4-methyl-5-vinyl, vinylsulfurol, vinyl sulfurol, fema no. 3313, 5-vinyl-4-methylthiazole, 4-methyl-5-vinyl-thiazole
PubChem CID 15654
IUPAC Name 5-ethenyl-4-methyl-1,3-thiazole
SMILES CC1=C(SC=N1)C=C

Specifications

Melting Point -15°C
Color Yellow
Boiling Point 66°C
Quantity 5 g
Formula Weight 125.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-5-vinylthiazole
RTECSNumber XJ5104000
TSCA Yes