missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-4H-thieno[3,2-b]pyrrol-5-yl)methanol, 97%, Thermo Scientific™

$152.35 - $152.35

Chemical Identifiers

CAS 121933-59-3
Molecular Formula C8H9NOS
Molecular Weight (g/mol) 167.23
MDL Number MFCD07772838
InChI Key ZRWPUHUHJRQLCD-UHFFFAOYSA-N
Synonym 4-methyl-4h-thieno 3,2-b pyrrol-5-yl methanol, 4-methylthieno 3,2-b pyrrol-5-yl methanol, 4h-thieno 3,2-b pyrrole-5-methanol,4-methyl, acmc-1c2v7, 4-methyl-4h-thieno 3,2-b pyrrol-5-yl-methanol, 4-methylthiopheno 2,3-d pyrrol-5-yl methan-1-ol
PubChem CID 6483918
IUPAC Name {4-methyl-4H-thieno[3,2-b]pyrrol-5-yl}methanol
SMILES CN1C(CO)=CC2=C1C=CS2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC40909DA
SDS View Documents
Thermo Scientific Chemicals
CC40909DA
1 g Amber Glass Bottle
Each for $152.35
Only null left
 
Description

Description

Specifications

Chemical Identifiers

121933-59-3
167.23
ZRWPUHUHJRQLCD-UHFFFAOYSA-N
6483918
CN1C(CO)=CC2=C1C=CS2
C8H9NOS
MFCD07772838
4-methyl-4h-thieno 3,2-b pyrrol-5-yl methanol, 4-methylthieno 3,2-b pyrrol-5-yl methanol, 4h-thieno 3,2-b pyrrole-5-methanol,4-methyl, acmc-1c2v7, 4-methyl-4h-thieno 3,2-b pyrrol-5-yl-methanol, 4-methylthiopheno 2,3-d pyrrol-5-yl methan-1-ol
{4-methyl-4H-thieno[3,2-b]pyrrol-5-yl}methanol

Specifications

121933-59-3
100.0
Amber Glass Bottle
MFCD07772838
4-methyl-4h-thieno 3,2-b pyrrol-5-yl methanol, 4-methylthieno 3,2-b pyrrol-5-yl methanol, 4h-thieno 3,2-b pyrrole-5-methanol,4-methyl, acmc-1c2v7, 4-methyl-4h-thieno 3,2-b pyrrol-5-yl-methanol, 4-methylthiopheno 2,3-d pyrrol-5-yl methan-1-ol
CN1C(CO)=CC2=C1C=CS2
167.23
167.23
(4-Methyl-4H-thieno[3, 2-b]pyrrol-5-yl)methanol
96.5
97%
C8H9NOS
1 g
ZRWPUHUHJRQLCD-UHFFFAOYSA-N
{4-methyl-4H-thieno[3,2-b]pyrrol-5-yl}methanol
6483918
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'