missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-4-heptanol 98.0+%, TCI America™
Supplier: TCI America M09545ML
Specifications
| 4-Methyl-4-heptanol | |
| Colorless | |
| C8H18O | |
| 5 mL | |
| IQXKGRKRIRMQCQ-UHFFFAOYSA-N | |
| 4-methylheptan-4-ol | |
| 136394 | |
| ≥98.0% (GC) |
| 598-01-6 | |
| 62°C | |
| MFCD00021840 | |
| 1987 | |
| CCCC(C)(CCC)O | |
| 130.231 | |
| 130.23 | |
| Liquid |
Chemical Identifiers
| 598-01-6 | |
| 130.231 | |
| IQXKGRKRIRMQCQ-UHFFFAOYSA-N | |
| 4-methylheptan-4-ol |
| C8H18O | |
| MFCD00021840 | |
| 136394 | |
| CCCC(C)(CCC)O |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : No