missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitroaniline, 97%, Thermo Scientific™

$47.33 - $422.59

Chemical Identifiers

CAS 119-32-4
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007910
InChI Key GDIIPKWHAQGCJF-UHFFFAOYSA-N
Synonym 4-amino-2-nitrotoluene, 3-nitro-4-methylaniline, 3-nitro-p-toluidine, m-nitro-p-toluidine, 2-nitro-4-aminotoluene, benzenamine, 4-methyl-3-nitro, 5-nitro-4-toluidine, p-toluidine, 3-nitro, gl-amin czech, gl-amin
PubChem CID 8390
ChEBI CHEBI:81670
IUPAC Name 4-methyl-3-nitroaniline
SMILES CC1=C(C=C(C=C1)N)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC127250050
View Documents Promotion Details
N/A 5g Glass bottle
Each for $47.33
Only null left
 
AC127250250
View Documents Promotion Details
N/A 25g Glass bottle
Each for $125.28
Only null left
 
AC127251000
View Documents Promotion Details
N/A 100g Glass bottle
Each for $422.59
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

119-32-4
152.15
GDIIPKWHAQGCJF-UHFFFAOYSA-N
8390
4-methyl-3-nitroaniline
C7H8N2O2
MFCD00007910
4-amino-2-nitrotoluene, 3-nitro-4-methylaniline, 3-nitro-p-toluidine, m-nitro-p-toluidine, 2-nitro-4-aminotoluene, benzenamine, 4-methyl-3-nitro, 5-nitro-4-toluidine, p-toluidine, 3-nitro, gl-amin czech, gl-amin
CHEBI:81670
CC1=C(C=C(C=C1)N)[N+](=O)[O-]

Specifications

119-32-4
100.0
96% min. (HPLC)
CH3C6H3(NO2)NH2
12, 996
GDIIPKWHAQGCJF-UHFFFAOYSA-N
4-methyl-3-nitroaniline
8390
152.15
4-Methyl-3-nitroaniline
96.0
Authentic
C7H8N2O2
MFCD00007910
4-amino-2-nitrotoluene, 3-nitro-4-methylaniline, 3-nitro-p-toluidine, m-nitro-p-toluidine, 2-nitro-4-aminotoluene, benzenamine, 4-methyl-3-nitro, 5-nitro-4-toluidine, p-toluidine, 3-nitro, gl-amin czech, gl-amin
CC1=C(C=C(C=C1)N)[N+](=O)[O-]
152.15
CHEBI:81670
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-314-

SDS
Documents

Documents

RUO – Research Use Only