missing translation for 'onlineSavingsMsg'
Learn More

4'-Methyl-3'-nitroacetophenone, 99%

Catalog No. AAL0937214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0937214 25 g
AAL0937206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0937214 Supplier Thermo Scientific Chemicals Supplier No. L0937214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5333-27-7 | C9H9NO3 | 179.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5333-27-7
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00033858
InChI Key YRBBMDOBWRUMEZ-UHFFFAOYSA-N
PubChem CID 79246
IUPAC Name 1-(4-methyl-3-nitrophenyl)ethanone
SMILES CC1=C(C=C(C=C1)C(=O)C)[N+](=O)[O-]

Specifications

Melting Point 58°C to 62°C
Quantity 25 g
Beilstein 1953352
Synonym 4'-methyl-3'-nitroacetophenone, 1-4-methyl-3-nitrophenyl ethanone, 4-methyl-3-nitroacetophenone, ethanone, 1-4-methyl-3-nitrophenyl, 1-4-methyl-3-nitrophenyl ethan-1-one, ethanone,1-4-methyl-3-nitrophenyl, 1-acetyl-4-methyl-3-nitrobenzene, pubchem3328, 3-nitro-4-methylacetophenon, ksc597m2h
Formula Weight 179.18
Percent Purity 99%
Chemical Name or Material 4'-Methyl-3'-nitroacetophenone
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only