missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-heptanol, erythro + threo, 99%

Catalog No. p-7025045
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2035706 5 g
AAB2035714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2035706 Supplier Thermo Scientific Chemicals Supplier No. B2035706
Only null left

CAS: 14979-39-6 | C8H18O | 130.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol, ether, acetone. Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 14979-39-6
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00004568
InChI Key BKQICAFAUMRYLZ-UHFFFAOYSA-N
Synonym 4-methyl-3-heptanol, 3-heptanol, 4-methyl, caswell no. 570a, epa pesticide chemical code 571200, 4-methyl-5-heptanol, acmc-1bqfj, dsstox_cid_21519, dsstox_rid_79763, dsstox_gsid_41519, 3-heptanol, 4-methyl-van8c
PubChem CID 26989
IUPAC Name 4-methylheptan-3-ol
SMILES CCCC(C)C(CC)O

Specifications

Density 0.83
Boiling Point 98°C to 99°C (75 mmHg)
Flash Point 59°C (138°F)
Refractive Index 1.431
Quantity 5 g
UN Number UN1987
Beilstein 1733008
Solubility Information Soluble in ethanol,ether,acetone. Slightly soluble in water.
Formula Weight 130.23
Percent Purity 99%
Chemical Name or Material 4-Methyl-3-heptanol, erythro + threo
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 239-058-9
RTECSNumber MJ4600000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only