missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylic acid, 97%, Thermo Scientific™

Catalog No. p-3522533
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC62001CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC62001CB Supplier Thermo Scientific Chemicals Supplier No. CC62001CB
Only null left

Chemical Identifiers

CAS 915707-58-3
Molecular Formula C9H10N2O3
Molecular Weight (g/mol) 194.19
MDL Number MFCD09065017
InChI Key JTDUYLQIBHWRRK-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-carboxylic acid, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazine-7-carboxylic acid, 2h-pyrido 3,2-b-1,4-oxazine-7-carboxylicacid, 3,4-dihydro-4-methyl, 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-carboxylicacid, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazine-7-carboxylic acid, 7-carboxy-3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazine, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b-1,4-oxazine-7-carboxylic acid, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazine-7-carboxylic acid
PubChem CID 24229645
IUPAC Name 4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazine-7-carboxylic acid
SMILES CN1CCOC2=C1N=CC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.