missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carbonitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC62016DE

 View more versions of this product

Catalog No. CC62016DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carbonitrile
96.5
97%
C9H9N3O
5g
VLSVLSBIHZDQTF-UHFFFAOYSA-N
4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazine-7-carbonitrile
24229651
97%
912569-63-2
100.0
Amber Glass Bottle
MFCD09065019
4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-carbonitrile, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazine-7-carbonitrile, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazine-7-carbonitrile, 2h-pyrido 3,2-b-1,4-oxazine-7-carbonitrile,3,4-dihydro-4-methyl
CN1CCOC2=C1N=CC(=C2)C#N
175.191
175.19

Chemical Identifiers

912569-63-2
175.191
VLSVLSBIHZDQTF-UHFFFAOYSA-N
24229651
CN1CCOC2=C1N=CC(=C2)C#N
C9H9N3O
MFCD09065019
4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-carbonitrile, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazine-7-carbonitrile, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazine-7-carbonitrile, 2h-pyrido 3,2-b-1,4-oxazine-7-carbonitrile,3,4-dihydro-4-methyl
4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazine-7-carbonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions