missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-2-thienyl)methylamine, 97%, Thermo Scientific™

Catalog No. p-3109907
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Catalog No. Quantity Packaging
CC30613CB 250 mg Amber Glass Bottle
1 options

Catalog No. CC30613CB

Supplier: Thermo Scientific Chemicals CC30613CB

Only null left

Chemical Identifiers

CAS 104163-39-5
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.205
MDL Number MFCD06657973
InChI Key CKQHNKAVFNDGMK-UHFFFAOYSA-N
Synonym 4-methylthiophen-2-yl methanamine, 4-methyl-2-thienyl methylamine, 2-thiophenemethanamine,4-methyl, 2-aminomethyl-4-methylthiophene, 4-methyl-2-aminomethyl thiophene, 4-methylthien-2-yl methylamine, 2-thiophenemethanamine, 4-methyl, 1-4-methylthiophen-2-yl methanamine, acmc-1cfbo, 4-methyl-2-thiophenemethanamine
PubChem CID 2795528
IUPAC Name (4-methylthiophen-2-yl)methanamine
SMILES CC1=CSC(=C1)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 127.21
Percent Purity 97%
Chemical Name or Material (4-Methyl-2-thienyl)methylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.