missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-thiazolecarboxaldehyde, 97%

Catalog No. AC429350010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429350010 1 g Glass bottle
AC429350050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC429350010 Supplier Thermo Scientific Chemicals Supplier No. 429350010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13750-68-0 | C5H5NOS | 127.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13750-68-0
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.17
MDL Number MFCD00022450
InChI Key NYMCQLLAIMUVSY-UHFFFAOYSA-N
Synonym 4-methylthiazole-2-carbaldehyde, 4-methyl-2-thiazolecarboxaldehyde, 4-methyl-thiazole-2-carbaldehyde, 4-methyl-1,3-thiazole-2-carboxaldehyde, 2-thiazolecarboxaldehyde, 4-methyl, 4-methylthiazole-2-carboxaldehyde, 4-methyl-2-formylthiazole, 2-formyl-4-methyl-1,3-thiazole, 4-methyl-2-thiazole carboxaldehyde, pubchem15253
PubChem CID 4263432
IUPAC Name 4-methyl-1,3-thiazole-2-carbaldehyde
SMILES CC1=CSC(=N1)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Boiling Point 90.0°C to 92.0°C (12.0 mbar)
Flash Point 67°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 127.17
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-thiazolecarboxaldehyde
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only