missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Quantity | 10g |
| Formula Weight | 214.22 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid |