missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid, 97%, Thermo Scientific™

Catalog No. CC20201DA
Change view
Click to view available options
Quantity:
1g
5g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC20201DA 1g
CC20201DE 5g
CC20201EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC20201DA Supplier Thermo Scientific Chemicals Supplier No. CC20201DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103249-79-2
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.224
InChI Key ZPXPNKCBUXSJFK-UHFFFAOYSA-N
Synonym 4-methyl-2-phenyl-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylicacid, 4-methyl-2-phenyl, acmc-1c8q0, 2-phenyl-4-methylpyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid,4-methyl-2-phenyl, 4-methyl-2-phenyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylic acid, 4-methyl-2-phenyl
PubChem CID 239975
IUPAC Name 4-methyl-2-phenylpyrimidine-5-carboxylic acid
SMILES CC1=NC(=NC=C1C(=O)O)C2=CC=CC=C2

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 214.22
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid