missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methanol, 97%, Thermo Scientific™
$109.03 - $109.03
Chemical Identifiers
| CAS | 61291-91-6 |
|---|---|
| Molecular Formula | C11H11NOS |
| Molecular Weight (g/mol) | 205.275 |
| MDL Number | MFCD01928782 |
| InChI Key | JSIPFWSPCWZJIU-UHFFFAOYSA-N |
| Synonym | 4-methyl-2-phenyl-1,3-thiazol-5-yl methanol, 4-methyl-2-phenylthiazol-5-yl methanol, 4-methyl-2-phenyl-thiazol-5-yl-methanol, 4-methyl-2-phenyl-1,3-thiazol-5-yl methan-1-ol, 4-methyl-2-phenyl-5-thiazolemethanol, 5-hydroxymethyl-4-methyl-2-phenylthiazole |
| PubChem CID | 736541 |
| IUPAC Name | (4-methyl-2-phenyl-1,3-thiazol-5-yl)methanol |
| SMILES | CC1=C(SC(=N1)C2=CC=CC=C2)CO |
Chemical Identifiers
| 61291-91-6 | |
| 205.275 | |
| JSIPFWSPCWZJIU-UHFFFAOYSA-N | |
| 736541 | |
| CC1=C(SC(=N1)C2=CC=CC=C2)CO |
| C11H11NOS | |
| MFCD01928782 | |
| 4-methyl-2-phenyl-1,3-thiazol-5-yl methanol, 4-methyl-2-phenylthiazol-5-yl methanol, 4-methyl-2-phenyl-thiazol-5-yl-methanol, 4-methyl-2-phenyl-1,3-thiazol-5-yl methan-1-ol, 4-methyl-2-phenyl-5-thiazolemethanol, 5-hydroxymethyl-4-methyl-2-phenylthiazole | |
| (4-methyl-2-phenyl-1,3-thiazol-5-yl)methanol |
Specifications
| 61291-91-6 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD01928782 | |
| 4-methyl-2-phenyl-1,3-thiazol-5-yl methanol, 4-methyl-2-phenylthiazol-5-yl methanol, 4-methyl-2-phenyl-thiazol-5-yl-methanol, 4-methyl-2-phenyl-1,3-thiazol-5-yl methan-1-ol, 4-methyl-2-phenyl-5-thiazolemethanol, 5-hydroxymethyl-4-methyl-2-phenylthiazole | |
| CC1=C(SC(=N1)C2=CC=CC=C2)CO | |
| 205.275 | |
| 205.28 | |
| (4-Methyl-2-phenyl-1, 3-thiazol-5-yl)methanol |
| 96.5 | |
| 97% | |
| C11H11NOS | |
| 1 g | |
| JSIPFWSPCWZJIU-UHFFFAOYSA-N | |
| (4-methyl-2-phenyl-1,3-thiazol-5-yl)methanol | |
| 736541 | |
| 97% |