missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenyl-1,3-oxazole-5-carbaldehyde, 95%, Thermo Scientific™

Catalog No. p-3629702
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC18104CB 250mg Amber Glass Bottle
CC18104DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC18104CB Supplier Thermo Scientific Chemicals Supplier No. CC18104CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 953408-85-0
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.198
MDL Number MFCD11109318
InChI Key NIYJTYPVRBYCEZ-UHFFFAOYSA-N
Synonym 5-formyl-4-methyl-2-phenyl-1,3-oxazole, 4-methyl-2-phenyloxazole-5-carbaldehyde, 4-methyl-2-phenyl-1,3-oxazole-5-carboxaldehyde
PubChem CID 26343583
IUPAC Name 4-methyl-2-phenyl-1,3-oxazole-5-carbaldehyde
SMILES CC1=C(OC(=N1)C2=CC=CC=C2)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 187.2
Percent Purity 95%
Chemical Name or Material 4-Methyl-2-phenyl-1, 3-oxazole-5-carbaldehyde