missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-phenyl-1,3-oxazole-5-carbaldehyde, 95%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 94.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 95% |
| Packaging | Amber Glass Bottle |
| Quantity | 1g |
| Formula Weight | 187.2 |
| Percent Purity | 95% |
| Chemical Name or Material | 4-Methyl-2-phenyl-1, 3-oxazole-5-carbaldehyde |