missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentenoic Acid (stabilized with HQ) 97.0+%, TCI America™

Catalog No. M02685ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02685ML 5 mL
M026825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M02685ML Supplier TCI America Supplier No. M02685ML

Chemical Identifiers

CAS 10321-71-8
Molecular Formula C6H9O2
Molecular Weight (g/mol) 113.14
MDL Number MFCD00043804
InChI Key QAOXMQCWUWZZNC-ONEGZZNKSA-M
Synonym 4-methyl-2-pentenoic acid, 4-methylpent-2-enoic acid, e-4-methylpent-2-enoic acid, 3-isopropyl acrylic acid, 2e-4-methylpent-2-enoic acid, 2-pentenoic acid, 4-methyl, unii-0qp1933ptd, 4-methylpent-2-en-1-oic acid, trans-4-methyl-2-pentenoic acid, e-4-methyl-2-pentenoic acid
PubChem CID 642039
IUPAC Name (2E)-4-methylpent-2-enoate
SMILES CC(C)\C=C\C([O-])=O

Specifications

Color Yellow
Boiling Point 212°C
Quantity 5 mL
UN Number 3265
Formula Weight 114.14
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentenoic Acid (stabilized with HQ)
EINECSNumber (2)-0609
TSCA Yes
Recommended Storage Refrigerator