missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-pentenoic Acid (stabilized with HQ) 97.0+%, TCI America™
Supplier: TCI America M026825ML
| Quantity | 25 mL |
|---|
Chemical Identifiers
| 10321-71-8 | |
| 113.14 | |
| QAOXMQCWUWZZNC-ONEGZZNKSA-M | |
| 642039 | |
| CC(C)\C=C\C([O-])=O |
| C6H9O2 | |
| MFCD00043804 | |
| 4-methyl-2-pentenoic acid, 4-methylpent-2-enoic acid, e-4-methylpent-2-enoic acid, 3-isopropyl acrylic acid, 2e-4-methylpent-2-enoic acid, 2-pentenoic acid, 4-methyl, unii-0qp1933ptd, 4-methylpent-2-en-1-oic acid, trans-4-methyl-2-pentenoic acid, e-4-methyl-2-pentenoic acid | |
| (2E)-4-methylpent-2-enoate |
Specifications
| 4-Methyl-2-pentenoic Acid (stabilized with HQ) | |
| Yellow | |
| C6H9O2 | |
| MFCD00043804 | |
| 4-methyl-2-pentenoic acid, 4-methylpent-2-enoic acid, e-4-methylpent-2-enoic acid, 3-isopropyl acrylic acid, 2e-4-methylpent-2-enoic acid, 2-pentenoic acid, 4-methyl, unii-0qp1933ptd, 4-methylpent-2-en-1-oic acid, trans-4-methyl-2-pentenoic acid, e-4-methyl-2-pentenoic acid | |
| CC(C)\C=C\C([O-])=O | |
| 113.14 | |
| 114.14 | |
| Liquid |
| 10321-71-8 | |
| 212°C | |
| 25 mL | |
| 3265 | |
| QAOXMQCWUWZZNC-ONEGZZNKSA-M | |
| (2E)-4-methylpent-2-enoate | |
| 642039 | |
| ≥97.0% (GC,T) |
Safety and Handling
EINECSNumber : (2)-0609
TSCA : Yes
Recommended Storage : Refrigerator