Learn More
4-Methyl-2-pentenoic acid, 98+%, Thermo Scientific Chemicals
$90.52 - $288.14
Chemical Identifiers
CAS | 10321-71-8 |
---|---|
Molecular Formula | C6H9O2 |
Molecular Weight (g/mol) | 113.14 |
MDL Number | MFCD00043804 |
InChI Key | QAOXMQCWUWZZNC-ONEGZZNKSA-M |
Synonym | 4-methyl-2-pentenoic acid, 4-methylpent-2-enoic acid, e-4-methylpent-2-enoic acid, 3-isopropyl acrylic acid, 2e-4-methylpent-2-enoic acid, 2-pentenoic acid, 4-methyl, unii-0qp1933ptd, 4-methylpent-2-en-1-oic acid, trans-4-methyl-2-pentenoic acid, e-4-methyl-2-pentenoic acid |
PubChem CID | 642039 |
IUPAC Name | (E)-4-methylpent-2-enoic acid |
SMILES | CC(C)\C=C\C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2263909
|
Thermo Scientific Chemicals
B2263909 |
10 g |
Each for $90.52
|
|
|||||
AAB2263918
|
Thermo Scientific Chemicals
B2263918 |
50 g |
Each for $288.14
|
|
|||||
Description
4-Methyl-2-pentenoic acid is used in medical research and in chemicals synthesis. It is used as a flavoring agents in foods & cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methyl-2-pentenoic acid is used in medical research and in chemicals synthesis. It is used as a flavoring agents in foods & cosmetics.
Solubility
Slightly soluble in water. Soluble in acetone, ether, ethanol.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Chemical Identifiers
10321-71-8 | |
113.14 | |
QAOXMQCWUWZZNC-ONEGZZNKSA-M | |
642039 | |
CC(C)\C=C\C([O-])=O |
C6H9O2 | |
MFCD00043804 | |
4-methyl-2-pentenoic acid, 4-methylpent-2-enoic acid, e-4-methylpent-2-enoic acid, 3-isopropyl acrylic acid, 2e-4-methylpent-2-enoic acid, 2-pentenoic acid, 4-methyl, unii-0qp1933ptd, 4-methylpent-2-en-1-oic acid, trans-4-methyl-2-pentenoic acid, e-4-methyl-2-pentenoic acid | |
(E)-4-methylpent-2-enoic acid |
Specifications
10321-71-8 | |
212°C | |
Unpleasant | |
1.4485 | |
MFCD00043804 | |
UN3265 | |
Slightly soluble in water. Soluble in acetone,ether,ethanol. | |
CC(C)\C=C\C([O-])=O | |
113.14 | |
114.15 | |
4-Methyl-2-pentenoic acid |
0.959 | |
>100°C (212°F) | |
C6H9O2 | |
(CH3)2CHCH=CHCO2H | |
10 g | |
4-methyl-2-pentenoic acid, 4-methylpent-2-enoic acid, e-4-methylpent-2-enoic acid, 3-isopropyl acrylic acid, 2e-4-methylpent-2-enoic acid, 2-pentenoic acid, 4-methyl, unii-0qp1933ptd, 4-methylpent-2-en-1-oic acid, trans-4-methyl-2-pentenoic acid, e-4-methyl-2-pentenoic acid | |
QAOXMQCWUWZZNC-ONEGZZNKSA-M | |
(E)-4-methylpent-2-enoic acid | |
642039 | |
≥98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 233-706-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only