missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentenoic acid, 98+%

Catalog No. AAB2263909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2263909 10 g
AAB2263918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2263909 Supplier Thermo Scientific Chemicals Supplier No. B2263909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10321-71-8 | C6H9O2 | 113.14 g/mol

4-Methyl-2-pentenoic acid is used in medical research and in chemicals synthesis. It is used as a flavoring agents in foods & cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-2-pentenoic acid is used in medical research and in chemicals synthesis. It is used as a flavoring agents in foods & cosmetics.

Solubility
Slightly soluble in water. Soluble in acetone, ether, ethanol.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 10321-71-8
Molecular Formula C6H9O2
Molecular Weight (g/mol) 113.14
MDL Number MFCD00043804
InChI Key QAOXMQCWUWZZNC-ONEGZZNKSA-M
Synonym 4-methyl-2-pentenoic acid, 4-methylpent-2-enoic acid, e-4-methylpent-2-enoic acid, 3-isopropyl acrylic acid, 2e-4-methylpent-2-enoic acid, 2-pentenoic acid, 4-methyl, unii-0qp1933ptd, 4-methylpent-2-en-1-oic acid, trans-4-methyl-2-pentenoic acid, e-4-methyl-2-pentenoic acid
PubChem CID 642039
IUPAC Name (E)-4-methylpent-2-enoic acid
SMILES CC(C)\C=C\C([O-])=O

Specifications

Density 0.959
Boiling Point 212°C
Flash Point >100°C (212°F)
Odor Unpleasant
Refractive Index 1.4485
Linear Formula (CH3)2CHCH=CHCO2H
Quantity 10 g
UN Number UN3265
Solubility Information Slightly soluble in water. Soluble in acetone,ether,ethanol.
Formula Weight 114.15
Percent Purity ≥98%
Chemical Name or Material 4-Methyl-2-pentenoic acid
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 233-706-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only