missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentenal, 95%

Catalog No. AC298350250
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC298350250 25 mL Glass bottle
AC298351000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC298350250 Supplier Thermo Scientific Chemicals Supplier No. 298350250

CAS: 5362-56-1 | C6H10O | 98.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5362-56-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
MDL Number MFCD00800501
InChI Key RIWPMNBTULNXOH-ONEGZZNKSA-N
Synonym 4-methyl-2-pentenal, unii-sxg1guk91a, e-4-methyl-2-pentenal, sxg1guk91a, fema no. 3510, 2-pentenal, 4-methyl, 4-methylpent-2-enal, 4-methyl-2-penten-1-al, 2-pentenal, 4-methyl-, 2e, 2e-4-methylpent-2-enal
PubChem CID 5916154
IUPAC Name (E)-4-methylpent-2-enal
SMILES CC(C)C=CC=O

Specifications

Acidity 15mg KOH/g max.
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8440g/mL
Boiling Point 70.0°C (75.0 mmHg)
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(CH3)CH=CHCHO
Refractive Index 1.4360 to 1.4440
Quantity 25 mL
Specific Gravity 0.844
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,propylene glycol,essential,oils
Formula Weight 98.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentenal
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
EINECSNumber 226-348-5

RUO – Research Use Only