Learn More
4-Methyl-2-pentenal, 95%
CAS: 5362-56-1 | C6H10O | 98.14 g/mol
$333.24 - $333.24
Chemical Identifiers
| CAS | 5362-56-1 |
|---|---|
| Molecular Formula | C6H10O |
| Molecular Weight (g/mol) | 98.14 |
| MDL Number | MFCD00800501 |
| InChI Key | RIWPMNBTULNXOH-ONEGZZNKSA-N |
| Synonym | 4-methyl-2-pentenal, unii-sxg1guk91a, e-4-methyl-2-pentenal, sxg1guk91a, fema no. 3510, 2-pentenal, 4-methyl, 4-methylpent-2-enal, 4-methyl-2-penten-1-al, 2-pentenal, 4-methyl-, 2e, 2e-4-methylpent-2-enal |
| PubChem CID | 5916154 |
| IUPAC Name | (E)-4-methylpent-2-enal |
| SMILES | CC(C)C=CC=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC298350250
|
Thermo Scientific Chemicals
298350250 |
25 mL | Glass bottle |
Each for $333.24
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5362-56-1 | |
| 98.14 | |
| RIWPMNBTULNXOH-ONEGZZNKSA-N | |
| 5916154 | |
| CC(C)C=CC=O |
| C6H10O | |
| MFCD00800501 | |
| 4-methyl-2-pentenal, unii-sxg1guk91a, e-4-methyl-2-pentenal, sxg1guk91a, fema no. 3510, 2-pentenal, 4-methyl, 4-methylpent-2-enal, 4-methyl-2-penten-1-al, 2-pentenal, 4-methyl-, 2e, 2e-4-methylpent-2-enal | |
| (E)-4-methylpent-2-enal |
Specifications
| 5362-56-1 | |
| 94.0 | |
| Colorless to Yellow | |
| 70.0°C (75.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4360 to 1.4440 | |
| MFCD00800501 | |
| 0.844 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol,propylene glycol,essential,oils | |
| CC(C)C=CC=O | |
| 98.14 | |
| 98.14 | |
| Liquid |
| 15mg KOH/g max. | |
| 100.0 | |
| 0.8440g/mL | |
| 32°C | |
| 94% min. (GC) | |
| C6H10O | |
| CH3CH(CH3)CH=CHCHO | |
| 25 mL | |
| 4-methyl-2-pentenal, unii-sxg1guk91a, e-4-methyl-2-pentenal, sxg1guk91a, fema no. 3510, 2-pentenal, 4-methyl, 4-methylpent-2-enal, 4-methyl-2-penten-1-al, 2-pentenal, 4-methyl-, 2e, 2e-4-methylpent-2-enal | |
| RIWPMNBTULNXOH-ONEGZZNKSA-N | |
| (E)-4-methylpent-2-enal | |
| 5916154 | |
| 95% | |
| 4-Methyl-2-pentenal |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
GHS Signal Word: Warning
EINECSNumber : 226-348-5
RUO – Research Use Only