missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, puriss. p.a., ACS Reagent, ≥99%, Honeywell Riedel-de Haën™

Puriss. p.a., ACS Reagent, Reag. Ph. Eur., for extraction analysis,≥99%

Supplier:  Honeywell Chemicals 334634X25L

 View more versions of this product

Catalog No. 60046262


Only null left
Explore available promotions

Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Specifications

4-Methyl-2-pentanone
108-10-1
Colorless
117°C
Characteristic
C6H12O
MFCD00008938
UN1245
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
CC(C)CC(=O)C
100.161
CHEBI:82344
≥99%
For extraction analysis
-80°C
0.801g/mL at 25°C
14°C (57.2°F)
Glass Bottle
(CH3)2CHCH2COCH3
4 x 2.5 L
605399
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methylpentan-2-one
7909
100.16g/mol
ACS/Reagent Ph. Eur./puriss. p.a.

Chemical Identifiers

108-10-1
100.161
NTIZESTWPVYFNL-UHFFFAOYSA-N
7909
4-methylpentan-2-one
C6H12O
MFCD00008938
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
CHEBI:82344
CC(C)CC(=O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P261-P305 + P351 + P338

DOTInformation : Class 3 - PG 2 - Methyl isobutyl ketone

RTECSNumber : 203-550-1

SDS
Documents

Documents

Promotions

Promotions